Learn More
Sodium tellurite(IV), 97%
Product Code. 11487527
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
Product Code. Quantity unitSize
11487527 50 g 50g
11497527 250 g 250g
2 options
Product Code. 11487527
Brand: Thermo Scientific Alfa Aesar A15633.18
Request Bulk or Custom Format
CAS: 10102-20-2 | H10Na2O9Te | 327.65 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10102-20-2
Molecular Formula H10Na2O9Te
Molecular Weight (g/mol) 327.65
MDL Number MFCD00036141
InChI Key OVBVNEXDNNHXIU-UHFFFAOYSA-L
Synonym sodium tellurite, disodium tellurite, sodium tellurate iv, disodium trioxotellurate, unii-0bb57la23y, tellurous acid, disodium salt, sodium tellurite iv, telluric acid h2teo3 , disodium salt, telluric acid, disodium salt
PubChem CID 24935
IUPAC Name disodium;tellurite
SMILES O.O.O.O.O.[Na+].[Na+].[O-][Te]([O-])(=O)=O
Specifications
Quantity 50 g
Linear Formula Na2TeO3
UN Number UN3284
Merck Index 14,8685
Formula Weight 221.58
Percent Purity 97%
Chemical Name or Material Sodium tellurite(IV)