Learn More
Sodium Sulfate Decahydrate, Supelco™
Product Code. 15854890
Change view
Click to view available options
Quantity:
1 kg
25 kg
Unit Size:
1kg
25kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15854890 1 kg 1kg
15827510 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15854890 Supplier Supelco Supplier No. 1066481000
 Restricted Item
Estimated Shipment: 26-11-2026
For analysis EMSURE™ ACS,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7727-73-3
Specifications
Melting Point 32.4°C
Quantity 1 kg
MDL Number MFCD00149189
Synonym Sodium sulfate decahydrate, Sodium sulfate decahydrate
Molecular Weight (g/mol) 322.2
Grade Research
Density 1.464 g/cm3
Chemical Name or Material Sodium Sulfate Decahydrate