missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfate, ≥99.0%, Reagent Grade, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 g
2.5 kg
5 kg
10 kg
Packaging:
Fiber drum
Poly bottle
Unit Size:
10kg
2.5kg
500g
5kg
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| Melting Point | 884°C |
| Color | Colorless |
| Physical Form | Solid |
| Quantity | 5 kg |
| Linear Formula | Na2SO4 |
| UN Number | NONH for all modes of transport |
| Formula Weight | 142.04g/mol |
| Percent Purity | ≥99.0% |
| Odor | Odorless |
| Packaging | Fiber drum |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction