missing translation for 'onlineSavingsMsg'
Learn More

Sodium solution for ICP 10g/L H2O matrix, Fisher chemical™

Catalog Number 12956344
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 10±0.02g/L
Packaging Bottle
Quantity 100 mL
Type Mass Spectrometry Standard
Analyte or Component Names Na
Solvent or Matrix H2O
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'