missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phthalate (Neutral), MP Biomedicals

Product Code. 11448594 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448594 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11448594 Supplier MP Biomedicals Supplier No. 0521593480

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 15968-01-1
Molecular Formula C8H4Na2O4
Quantity 100 g
Synonym disodium phthalate, sodium phthalate, phthalic acid disodium salt, unii-qb14yv193c, 1,2-benzenedicarboxylic acid, disodium salt, sodium 2-carboxybenzoate, 1,2-benzenedicarboxylic acid, sodium salt 1:2, 88-99-3 parent, disodiumphthalate, acmc-209dk3
InChI Key HQWKKEIVHQXCPI-UHFFFAOYSA-L
SMILES C1=CC=C(C(=C1)C(=O)[O-])C(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;phthalate
Molecular Weight (g/mol) 210.096
PubChem CID 85215
Chemical Name or Material Sodium Phthalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.