missing translation for 'onlineSavingsMsg'
Learn More

Melford Sodium Phosphate Dibasic, Anhydrous

Product Code. 30018784
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018784 5 kg 5kg
30018783 500 g 500g
30018782 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018784 Supplier Melford Supplier No. S231005000.0
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sodium Phosphate Dibasic Anhydrous is a chemical compound that is commonly known as disodium hydrogen phosphate or disodium phosphate.

This compound is an anhydrous form of sodium phosphate, meaning it does not contain any water molecules.

In the life science field, Sodium Phosphate Dibasic Anhydrous is primarily used as a buffering agent, which helps maintain a stable pH in various biological and biochemical processes.

It is often utilized in laboratory settings for the preparation of various biological reagents, cell culture media, and protein purification processes. The compound's buffering properties make it valuable in the formulation of diagnostic kits and pharmaceutical products, where maintaining a specific pH is critical for the stability and effectiveness of the final product

Specifications

Chemical Name or Material Sodium Phosphate Dibasic, Anhydrous
CAS 7558-79-4
Quantity 5 kg
Molecular Formula Na2HPO4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.