Learn More
Sodium Phenoxide Trihydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Sodium Phenoxide Trihydrate |
| CAS | 156150-40-2 |
| Molecular Formula | C6H5NaO.3[H2O] |
| Quantity | 100 g |
| Synonym | Sodium Salt Phenol Hydrate (1:1:3), Phenol Sodium Salt Trihydrate |
| InChI Formula | InChI=1S/C6H6O.Na.3H2O/c7-6-4-2-1-3-5-6;;;;/h1-5,7H;;3*1H2/q;+1;;;/p-1 |
| SMILES | O.O.O.[Na+].[O-]c1ccccc1 |
| IUPAC Name | sodium;phenoxide;trihydrate |
| Molecular Weight (g/mol) | 170.14 |
| Formula Weight | 170.0555 |