missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phenolate Trihydrate, Sigma-Aldrich™

Product Code. 15881221
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15881221 500 g 500g
15871221 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15881221 Supplier Sigma Aldrich Supplier No. 8208730500
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156150-40-2

Specifications

Melting Point 56°C to 61°C
MDL Number MFCD00149636
Quantity 500 g
Synonym Sodium phenolate trihydrate, Phenol sodium salt, Sodium phenolate trihydrate
Molecular Weight (g/mol) 170.14
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.