missing translation for 'onlineSavingsMsg'
Learn More

Sodium peroxydisulfate, 98%

Product Code. p-4407926
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 11448660

Brand: Thermo Scientific Alfa Aesar 054100.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7775-27-1 | Na2O8S2 | 238.092 g/mol

Sodium peroxydisulfate is used as a radical initiator for emulsion polymerization reactions like acrylonitrile butadiene styrene, detergent component, soil conditioner and soil remediation. It is also used for curing of formaldehyde adhesives. It acts as a bleaching agent and in the production of dyestuffs. It finds application in zinc and printed circuit boards and considered to be a replacement for ammonium persulfate in etching mixtures. Further, it is used in the preparation of diapocynin from apocynin. It plays a role in the conversion of phenols to para-diphenols in alkaline solution and of arylamines to aminophenols. It is actively involved in the Elbs persulfate oxidation and the Boyland-Sims oxidation reactions as an oxidizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-27-1
Molecular Formula Na2O8S2
Molecular Weight (g/mol) 238.092
MDL Number MFCD00003501
InChI Key CHQMHPLRPQMAMX-UHFFFAOYSA-L
Synonym sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate
PubChem CID 62655
IUPAC Name disodium;sulfonatooxy sulfate
SMILES [O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+]

Specifications

Physical Form Crystalline
Quantity 1 kg
Assay Percent Range 98%
Solubility Information Soluble in water.
Formula Weight 238.1
Density 2.40 g/mL
Chemical Name or Material Sodium peroxydisulfate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium peroxydisulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Oxidising solid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contains : Sodium persulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.