missing translation for 'onlineSavingsMsg'
Learn More

Sodium perborate tetrahydrate, MilliporeSigma™

Product Code. 15874860
Change view
Click to view available options
Quantity:
50 kg
Packaging:
Fibre Carton
Unit Size:
50kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15874860 50 kg Fibre Carton 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15874860 Supplier Supelco Supplier No. 1065609050
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity sodium perborate tetrahydrate suitable for a variety of laboratory applications.

TRUSTED_SUSTAINABILITY

Specifications

CAS 10486-00-7
Physical Form Powder
Quantity 50 kg
Molecular Formula BNaO3 · 4H2O
Linear Formula NaBO3 · 4H2O
Solubility Information 23g/L (20°C)
Molecular Weight (g/mol) 153.86
Formula Weight 153.88
Grade Pure
Packaging Fibre Carton
Density 1.73g/cm3; Bulk Density: 800 kg/m3
Chemical Name or Material Sodium Perborate Tetrahydrate, Sodium perborate tetrahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.