missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitroprusside Dihydrate, (Crystalline),MP Biomedicals

Product Code. 11476105 Shop All MP Biomedicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
250 g
500 g
missing translation for 'unitSize'
100g
250g
500g
This item is not returnable. View return policy

Product Code. 11476105

missing translation for 'mfr': MP Biomedicals 0215206183

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reagent used for chromatographic detection of peptides containing cystine. Also liberates nitric oxide in biological systems

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13755-38-9
Molecular Formula C5H4FeN6Na2O3
Molecular Weight (g/mol) 297.95
MDL Number MFCD00149192
InChI Key JFPDVRGGFBUORC-UHFFFAOYSA-N
Synonym nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
PubChem CID 11953895
SMILES O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]

Specifications

Physical Form Crystalline
Quantity 250 g
IUPAC Name disodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate
Formula Weight 297.95
Chemical Name or Material Sodium Nitroprusside Dihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.