Learn More
Sodium nitrite, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E250, 99 to 100.5% (calc. to the dried substance), Solstice
BIOPHARMA
Product Code. 15651330
Change view
Click to view available options
Quantity:
1 kg
5 kg
50 kg
Unit Size:
1kg
50kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15651330 50 kg 50kg
15641330 1 kg 1kg
15661330 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15651330 Supplier Honeywell-Fluka Supplier No. 1344750KG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E250, 99-100.5% (calc. to the dried substance)
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium;nitrite
SMILES [Na+].[O-]N=O
Specifications
Melting Point 271°C (literature)
CAS Max % 100.0000%
Quantity 50 kg
Assay Percent Range 99 to 100.5%
Linear Formula NaNO2
UN Number UN1500
Formula Weight 69
Percent Purity 99 to 100.5% (calc. to the dried substance)
Grade puriss.
Ignition Point 914°F
Packaging Fiberboard Box
Density 2.17 g/cm3
Chemical Name or Material Sodium nitrite, Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E250, 99-100.5% (calc. to the dried substance)