missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite, For Analysis, EMSURE™, MilliporeSigma™

Product Code. 15834860
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15834860 500 g Plastic Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15834860 Supplier Supelco Supplier No. 1065490500
 Restricted Item
Estimated Shipment: 22-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recommended Storage

+5°C to +30°C.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 280°C (decomposition)
CAS 7632-00-0
Color White
pH 9 (100g/L, H2O, 20°C)
Physical Form Solid
Quantity 500 g
Molecular Formula NNaO2
MDL Number MFCD00011118
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
Solubility Information 820g/L
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
SMILES [Na+].[O-]N=O
IUPAC Name sodium nitrite
Molecular Weight (g/mol) 69.00
PubChem CID 23668193
ChEBI CHEBI:78870
Formula Weight 69
Percent Purity >99.0%
Packaging Plastic Bottle
Density 2.1 g/3 (20 °C)
Chemical Name or Material Sodium Nitrite
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.