missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitrite, ACS Reagent, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
5g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 5 g |
| Linear Formula | NaNO2 |
| UN Number | 1500 |
| Formula Weight | 69g/mol |
| Percent Purity | ≥97% |
| Grade | ACS Reagent |
| Ignition Point | 914°F |
| Chemical Name or Material | Sodium Nitrite |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction