Learn More
Sodium nitrate, Scharlab
Product Code. 11318554
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318554 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11318554 Supplier Scharlab Supplier No. SO05010500
  • ONLINE EXCLUSIVE PRICE
    451.50 NOK
    427.34 NOK / 500g
    Save 24.16 NOK
    5% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Specifications
CAS 7631-99-4
Quantity 500 g
Molecular Formula NNaO3
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
SMILES [Na+].[O-][N+]([O-])=O
IUPAC Name sodium nitrate
Molecular Weight (g/mol) 84.99
Grade Reagent
compliance-icons compliance-icons
Product Identifier
  • Sodium nitrate, reagent grade, ACS, ISO
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to .