missing translation for 'onlineSavingsMsg'
Learn More

Sodium Molybdate Dihydrate, 99.99% Trace Metals Basis, Honeywell™

Product Code. 15655650 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly Bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15655650

Brand: Honeywell Research Chemicals 480967100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

99.99% trace metals basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-40-6
Molecular Formula H4MoNa2O6
Molecular Weight (g/mol) 241.96
MDL Number MFCD00149170
InChI Key FDEIWTXVNPKYDL-UHFFFAOYSA-N
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
PubChem CID 16211258
ChEBI CHEBI:75213
IUPAC Name disodium dioxomolybdenumbis(olate) dihydrate
SMILES O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O

Specifications

Melting Point 100°C
Color White
pH 7 to 10 (at 20°C)
Physical Form Liquid
Quantity 100 g
Linear Formula Na2MoO4 · 2H2O
UN Number NONH for all modes of transport
Formula Weight 241.95g/mol
Percent Purity 99.99% (trace metals basis)
Odor Odorless
Packaging Poly Bottle
Density 2.37g/mL at 25°C
Chemical Name or Material Sodium molybdate dihydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.