Learn More
Sodium Methanesulfonate-d3, CDN
Specifications
Specifications
| Chemical Name or Material | Methanesulfonic Acid-D3 Sodium Salt |
| CAS | 1610056-71-7 |
| Molecular Formula | C D3 O3 S . Na |
| Quantity | 1 g |
| Synonym | Methane-d3-sulfonic acid, sodium salt (1:1) (ACI), Methanesulfonic acid-D3 sodium salt, Sodium mesylate-d3, Sodium methanesulfonate-d3 |
| InChI Formula | InChI=1 S/CH4O3S.Na/c1-5(2,3)4;/h1H3,(H,2,3,4);/q;+1/p-1/i1D3; |
| SMILES | [Na+].[2 H]C([2 H])([2 H])S(=O)(=O)[O-] |
| IUPAC Name | sodium;trideuteriomethanesulfonate |
| Molecular Weight (g/mol) | 121.106 |
| Formula Weight | 120.9889 g/mol |
| Show More |