Learn More
Sodium methanesulfonate, 98%
Product Code. p-7026441
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400587 5 g 5g
11410587 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11400587 Supplier Thermo Scientific Alfa Aesar Supplier No. B22439.06
Request Bulk or Custom Format
CAS: 2386-57-4 | CH3NaO3S | 118.082 g/mol
Sodium methanesulfonate is used as a non-steroidal anti-inflammatory drug.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium methanesulfonate is used as a non-steroidal anti-inflammatory drug.
Solubility
Soluble in water, alcohol, ether, hexane, benzene, methylcyclopentane, toluene, o-chlorotoluene and ethyl disulfide.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2386-57-4
Molecular Formula CH3NaO3S
Molecular Weight (g/mol) 118.082
MDL Number MFCD00064389
InChI Key KKVTYAVXTDIPAP-UHFFFAOYSA-M
Synonym sodium methanesulfonate, methanesulfonic acid sodium salt, methanesulfonic acid, sodium salt, sodium methanesulphonate, sodium mesylate, methylsulfonic acid sodium salt, 75-75-2 parent, sodium methyl sulfonate, methanesulfonic acid, sodium salt 1:1, sodium methylsulfonate
PubChem CID 638112
IUPAC Name sodium;methanesulfonate
SMILES CS(=O)(=O)[O-].[Na+]
Specifications
Odor Odorless
Linear Formula CH3SO3Na
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water,alcohol,ether,hexane,benzene,methylcyclopentane,toluene,o-chlorotoluene and ethyl disulfide.
Formula Weight 118.09
Percent Purity 98%
Chemical Name or Material Sodium methanesulfonate