Learn More
Sodium Iodide, Supelco™
Specifications
Specifications
| Boiling Point | 1304°C/1013 hPa |
| Melting Point | 659°C |
| CAS | 7681-82-5 |
| Quantity | 100 g |
| MDL Number | MFCD00003532 |
| Synonym | Sodium iodide, Sodium iodide |
| Molecular Weight (g/mol) | 149.89 |
| Grade | Research |
| Density | 3.67 g/cm3 at 20°C |
| Chemical Name or Material | Sodium Iodide |