Learn More
Sodium Hydroxide Solution, 40% Volume, Chem-Lab
Product Code. 11938534
Click to view available options
Quantity:
5 L
Unit Size:
5 L
Product Code. 11938534
Brand: Chem-Lab CL02.1411.5000
Used as a volumetric solution in laboratory applications. Sodium Hydroxide Solution, 40% Volume, Chem-Lab is a strong base and is highly caustic. It is used in titration and to increase alkalinity, to neutralize acids and to make sodium salts. One liter of the solution contains 400g of NaOH.
Sodium hydroxide is used in many industries, mostly as a strong chemical base in the manufacture of pulp and paper, textiles, drinking water, soaps and detergents and as a drain cleaner.
Specifications
pH >13
Color Colorless
Boiling Point 100°C
CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Concentration 40% by volume
Density 1.329g/mL
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Water Soluble
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 5 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40.00g/mol
Concentration or Composition (by Analyte or Components) 400g NaOH per 1L H2O
Odor Odorless
Solvent or Matrix Water
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide 40 vol. % solution
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide