Learn More
Sodium Hydroxide, Ethanolic, Ph. Eur. Grade, 01.M, Reagecon™
BIOPHARMA
Product Code. 11494809 Shop All Reagecon Products
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Product Code. 11494809
Brand: Reagecon 3007000
Specifications
Concentration 0.1M
Quantity 1 L
compliance-icons compliance-icons
Product Identifier
  • 0.1M SODIUM HYDROXIDE, ETHANOLIC
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
  • MIXTURE LIST-Contains : ethyl alcohol