Learn More
Sodium Hydroxide, 5M, Chem Lab™
Product Code. 11903243
Change view
Click to view available options
Quantity:
5 L
10 L
Packaging:
473mL
10L
Unit Size:
10 L
5 L
Product Code. Quantity Packaging unitSize
11903243 5 L 473mL 5 L
11923253 10 L 10L 10 L
2 options
Product Code. 11903243
Brand: Chem-Lab CL05.1425.5000
Specifications
Color Colorless
Concentration 200g NaOH/l H2O = 5 N (±0.01/20°C)
Density 1.18g/mL
Solubility Information Soluble in water
Quantity 5 L
Odor Odourless
Packaging 473mL
Physical Form Liquid
compliance-icons
Product Identifier
  • Sodium hydroxide 5 mol/l
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide, pellets a.r.