Learn More
Sodium Hydroxide, 40% w/v, Extra Pure, Scharlab
Product Code. 11358524
Click to view available options
Quantity:
5 L
Packaging:
HDPE Carboy
Unit Size:
5 L
Product Code. 11358524
Brand: Scharlab SO0422005P
Suitable for qualitative and semi-quantitative work. Sodium Hydroxide, 40% w/v, Extra Pure, Scharlab is often used in organic chemistry for purifying synthesized products when higher grade solvents than Synthesis Grade are required.
Extra Pure grade specifications are less strict than Reagent Grade, but are significantly higher than industrial products. They are often used in the pharmaceutical industry, either as a reagent or as raw materials for production.
Specifications
Color Colorless
CAS 1310-73-2
Molecular Formula HNaO
UN Number 1824
Density 1.3293g/cm3
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Miscible in water
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 5 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40.00g/mol
Percent Purity ≥40%
Grade Applications: analytical chemistry, laboratory reagent, solvents, for pharmaceutical use, for the synthesis of inorganic salts
Packaging HDPE Carboy
Physical Form Solution
Assay w/v, acidimetric
Chemical Name or Material Sodium hydroxide
Show More Show Less
compliance-icons
Product Identifier
  • Sodium hydroxide, solution 40% w/v, extra pure
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Sodium Hydroxide