Learn More
Sodium Hydroxide, 0.1 mol/l in Ethanol, Chem Lab™
Product Code. 12536548
Click to view available options
:
Quantity: 1L; Density: 0.84g/mL
Product Code. 12536548
Brand: Chem-Lab CL07.1429.1000
Specifications
pH >11
Color Colorless
Concentration 4g NaOH/l C2H5OH = 0.1 N
Density 0.84g/mL
Solubility Information Soluble in Water
Quantity 1 L
Odor Ethanolic
Physical Form Liquid
compliance-icons
Product Identifier
  • Sodium hydroxide 0.1 mol/l in Ethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : Sodium hydroxide