missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydrogen Sulfate Monohydrate, Supelco™

Product Code. 15877450
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15877450 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15877450 Supplier Supelco Supplier No. 1063520500
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 183°C (Decomposition)
CAS 10034-88-5
Quantity 500 g
MDL Number MFCD00149210
Synonym Sodium hydrogen sulfate monohydrate, sodium bisulfate monohydrate, sodium bisulphate monohydrate, Sodium hydrogen sulfate monohydrate
Molecular Weight (g/mol) 120.06 (Anhydrous Basis)
Grade Research
Chemical Name or Material Sodium Hydrogen Sulfate Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.