Learn More
Sodium hexafluorophosphate, 98%
Product Code. 11449936
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449936 5 g 5g
11459936 25 g 25g
11469936 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11449936 Supplier Thermo Scientific Alfa Aesar Supplier No. B20898.06
Request Bulk or Custom Format
CAS: 21324-39-0 | F6NaP | 167.95 g/mol
Sodium hexafluorophosphate is used as an ion pairing agent for the determination of cobalt(II) in water samples. It is used as an important raw material in the preparation of 2-chloro-1,3-bis(dimethylamino)trimethinium hexafluorophosphate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium hexafluorophosphate is used as an ion pairing agent for the determination of cobalt(II) in water samples. It is used as an important raw material in the preparation of 2-chloro-1,3-bis(dimethylamino)trimethinium hexafluorophosphate.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong acids, bases and oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21324-39-0
Molecular Formula F6NaP
Molecular Weight (g/mol) 167.95
MDL Number MFCD00011122
InChI Key KMADQUOCJBLXRP-UHFFFAOYSA-N
Synonym sodium hexafluorophosphate, sodium hexa-fluoro phosphate, sodium hexafluorophosphate v, napf6, sodium hexaflurophosphate, acmc-1cao2, sodium hexafluoro-, e5-phosphanuide, sodium hexafluoro-$l^ 5-phosphanuide, phosphate 1-, hexafluoro-, sodium 1:1
PubChem CID 5147921
SMILES [Na+].F[P-](F)(F)(F)(F)F
Specifications
Melting Point >200°C
Quantity 5 g
UN Number UN3260
Solubility Information Soluble in water.
IUPAC Name sodium hexafluoro-λ⁵-phosphanuide
Formula Weight 167.95
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 2.37
Chemical Name or Material Sodium hexafluorophosphate