Learn More
Sodium hexachlororhodate(III) dodecahydrate, Rh 17.1%, Thermo Scientific Chemicals
Product Code. 11301858
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11301858 1 g 1g
11391848 5 g 5g
2 options
Product Code. 11301858
Brand: Thermo Scientific Alfa Aesar 011809.03
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14972-70-4
Molecular Formula Cl6H24Na3O12Rh
Molecular Weight (g/mol) 600.75
MDL Number MFCD00003509
InChI Key JCMYDKHYUGZPJG-UHFFFAOYSA-H
IUPAC Name trisodium hexachlororhodiumheptakis(ylium) dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].Cl[Rh+7](Cl)(Cl)(Cl)(Cl)Cl
Specifications
Melting Point 900°C (decomposition)
Quantity 1 g
Formula Weight 600.77 (384.59 Anhydrous)
Concentration or Composition (by Analyte or Components) Rh 17.1%
Physical Form Crystalline
Chemical Name or Material Sodium hexachlororhodate(III) dodecahydrate