Learn More
Sodium formononetin-3'-sulfonate, MedChemExpress
Product Code. 16465528
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16465528
Brand: Medchem Express HY13063
MedChemExpress Sodium formononetin-3'-sulfonate (Sul-F) is a water-sol.
Sodium formononetin-3'-sulfonate (Sul-F) is a water-sol.
Specifications
Chemical Name or Material Sodium formononetin-3'-sulfonate
CAS 949021-68-5
Color White
Purity Grade Notes Research
Molecular Formula C16H11NaO7S
Quantity 10 mg
Synonym Sul-F
Solubility Information H2O : 25 mg/mL (67.51 mM; Need ultrasonic)
SMILES OC1=CC=C(C2=C1)C(C(C3=CC(S(=O)(O[Na])=O)=C(OC)C=C3)=CO2)=O
Molecular Weight (g/mol) 370.31
Formula Weight 370.31
Percent Purity 99.79%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.