Learn More
Sodium dodecyl sulfate, 99.5%, MP Biomedicals™
Product Code. 11476571 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
Product Code. 11476571
Brand: MP Biomedicals 0219483191
Chemical Identifiers
CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym Sodium lauryl sulfate, SDS
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
Specifications
Color White
Physical Form Granular Powder
Quantity 1 kg
Formula Weight 288.38
Percent Purity 99.5%
Chemical Name or Material Sodium dodecyl sulfate