Learn More
Sodium Dichromate Dihydrate, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | Sodium Dichromate Dihydrate |
| CAS | 7789-12-0 |
| Molecular Formula | Cr2Na2O7.2(H2O) |
| Quantity | 50 g |
| Synonym | Dichromic Acid (H2Cr2O7) Disodium Salt Dihydrate, Dichromic Acid Disodium Salt Dihydrate, Disodium Dichromate Dihydrate, Sodium Chromate (Na2Cr2O7) Dihydrate, Sodium Dichromate (Na2Cr2O7) Dihydrate, Sodium Dichromate Dihydrate, Sodium Dichromate Dihydrate (Na2Cr2O7.2H2O) |
| InChI Formula | InChI=1S/2Cr.2Na.2H2O.7O/h;;;;2*1H2;;;;;;;/q;;2*+1;;;;;;;;2*-1 |
| SMILES | O.O.[Na+].[Na+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] |
| IUPAC Name | disodium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium;dihydrate |
| Molecular Weight (g/mol) | 261.97 |
| Formula Weight | 297.8461 |