Learn More
Sodium (dichloro(phosphono)methyl)phosphonate tetrahydrate, 99%, BLD Pharm™
Specifications
Specifications
| CAS | 88416-50-6 |
| Molecular Formula | CH10Cl2Na2O10P2 |
| MDL Number | MFCD02940846 |
| Quantity | 25 g |
| SMILES | ClC(P([O-])([O-])=O)(P(O)(O)=O)Cl.[H]O[H].[H]O[H].[H]O[H].[H]O[H].[Na+].[Na+] |
| Molecular Weight (g/mol) | 360.92 |
| Percent Purity | 99% |
| Chemical Name or Material | Sodium (dichloro(phosphono)methyl)phosphonate tetrahydrate |