Learn More
Sodium Diatrizoate, MP Biomedicals™
Product Code. p-7083107 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
Product Code. Quantity unitSize
11416302 10 g 10g
11426302 50 g 50g
11436302 100 g 100g
3 options
Product Code. 11416302
Brand: MP Biomedicals 0210290810
Used as a density gradient reagent in blood cell separation. Sodium Diatrizoate, MP Biomedicals is also utilized as a radiological contrast agent. It causes partial redistribution of tight-junction-associated membrane proteins into a cytoplasmic compartme
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 737-31-5
Molecular Formula C11H8I3N2NaO4
Molecular Weight (g/mol) 635.898
InChI Key ZEYOIOAKZLALAP-UHFFFAOYSA-M
Synonym diatrizoate sodium, sodium diatrizoate, sodium amidotrizoate, hypaque sodium, triombrin, triombrine, hypaque, vascoray, hypaque cysto, hpaque-cysto
PubChem CID 23672589
ChEBI CHEBI:53692
IUPAC Name sodium;3,5-diacetamido-2,4,6-triiodobenzoate
SMILES CC(=O)NC1=C(C(=C(C(=C1I)C(=O)[O-])I)NC(=O)C)I.[Na+]
Specifications
Physical Form Powder
Quantity 10 g
Solubility Information Soluble in water (350mg/mL).
Formula Weight 635.9
Percent Purity >99%