Learn More
VEE GEE Sodium Chloride Hydrometer
Product Code. 17886553
Click to view available options
Sodium Chloride Range:
0 to 100% by Saturation
0 to 26.5% by Weight
Unit Size:
Each
Product Code. 17886553
Brand: VEE GEE 66111
The Sodium Chloride Hydrometer measures the percentage of sodium chloride.
  • Calibration for use at 15.5°C/60°F
  • Approx. Length: 305 mm
Specifications
Type Salt Hydrometer
Increments 1.00%
Length (Metric) 30.5 cm
Temperature of Standardization 15.5°C (60°F)
Sodium Chloride Range 0 to 100% by Saturation