Learn More
VEE GEE Sodium Chloride Hydrometer
Click to view available options
Sodium Chloride Range:
0 to 100% by Saturation
0 to 26.5% by Weight
Unit Size:
Each
Description
- Calibration for use at 15.5°C/60°F
- Approx. Length: 305 mm
Specifications
Specifications
| Type | Salt Hydrometer |
| Increments | 0.50% |
| Length (Metric) | 30.5 cm |
| Temperature of Standardization | 15.5°C (60°F) |
| Sodium Chloride Range | 0 to 26.5% by Weight |
Product Content Correction
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu