Learn More
Sodium Butyrate, Sigma-Aldrich™
Product Code. 15810841
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15810841 1 kg 1kg
15800841 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15810841 Supplier Sigma Aldrich Supplier No. 8202361000
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 156-54-7
Specifications
Melting Point 244°C
Density 1.324 g/cm3 at 30°C
Quantity 1 kg
MDL Number MFCD00002816
Synonym Sodium butyrate, Butyric acid sodium salt, Sodium butyrate
Molecular Weight (g/mol) 110.09
Grade Ungraded
Chemical Name or Material Sodium Butyrate