Learn More
Sodium bromoacetate, 98%
Product Code. 11368096
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11368096 5 g 5g
11378096 25 g 25g
2 options
Product Code. 11368096
Brand: Thermo Scientific Alfa Aesar L14983.06
Request Bulk or Custom Format
CAS: 1068-52-6 | C2H2BrNaO2 | 160.93 g/mol
It is an important raw material and intermediate used in organic synthesis, agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1068-52-6
Molecular Formula C2H2BrNaO2
Molecular Weight (g/mol) 160.93
MDL Number MFCD01311832
InChI Key SESSOVUNEZQNBV-UHFFFAOYSA-M
Synonym sodium bromoacetate, sodium 2-bromoacetate, sodium monobromoacetate, unii-8j155ck8pv, acetic acid, bromo-, sodium salt, bromoacetic acid sodium salt, acmc-1bowb, sodium bromoacetate, ?brch2coona, acetic acid,bromo-,sodium salt 8ci,9ci, acetic acid, bromo-,sodium salt 8ci,9ci
PubChem CID 23675628
IUPAC Name sodium;2-bromoacetate
SMILES C(C(=O)[O-])Br.[Na+]
Specifications
Melting Point >300°C
Odor Odorless
Quantity 5 g
UN Number UN2811
Solubility Information Soluble in water.
Formula Weight 160.93
Percent Purity 98%
Chemical Name or Material Sodium bromoacetate