Learn More
Sodium Borohydride, 98-99%, (Crystalline), MP Biomedicals
Product Code. 11446282 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
50 g
100 g
500 g
Unit Size:
100g
25g
500g
50g
Product Code. 11446282
Brand: MP Biomedicals 0210289450
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Excellent reducing agent
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]
Specifications
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 98-99%
Formula Weight 37.8
Chemical Name or Material Sodium Borohydride