Learn More
Sodium Barbiturate, Sigma-Aldrich™
Product Code. 15880831
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15880831 250 g 250g
15870831 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15880831 Supplier Sigma Aldrich Supplier No. 8202330250
 Restricted Item
Estimated Shipment: 19-11-2026
≥97%
for synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4390-16-3
Specifications
Quantity 250 g
MDL Number MFCD00065377
Synonym Sodium barbiturate, Malonylurea natrium salt, Sodium barbiturate
Molecular Weight (g/mol) 150.07
Grade Ungraded
Chemical Name or Material Sodium Barbiturate