Learn More
Sodium 2,4-Dimethylbenzenesulfonate Monohydrate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 142063-30-7 |
| Molecular Formula | C8H11NaO4S |
| MDL Number | MFCD15071080 |
| Quantity | 100 g |
| SMILES | O=S(C1=CC=C(C)C=C1C)([O-])=O.[H]O[H].[Na+] |
| Molecular Weight (g/mol) | 226.23 |
| Percent Purity | 98% |
| Chemical Name or Material | Sodium 2,4-Dimethylbenzenesulfonate Monohydrate |