missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-tetradecyl sulfate, 95%, cont. up to ca 5% sodium methyl sulfate, Thermo Scientific Chemicals

Catalog Number p-7026123
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1191-50-0
C14H29NaO4S
316.43
MFCD00007468
UPUIQOIQVMNQAP-UHFFFAOYSA-M
sodium tetradecyl sulfate, sodium myristyl sulfate, niaproof 4, sodium tetradecyl sulphate, tesapon k 14, texapon k 14, trombavar, natri tetradecylsulfas, sotradecol sodium, tetradecyl sodium sulfate
23665770
sodium;tetradecyl sulfate
[Na+].CCCCCCCCCCCCCCOS([O-])(=O)=O
This item is not returnable. View return policy
1191-50-0
C14H29NaO4S
316.43
MFCD00007468
UPUIQOIQVMNQAP-UHFFFAOYSA-M
sodium tetradecyl sulfate, sodium myristyl sulfate, niaproof 4, sodium tetradecyl sulphate, tesapon k 14, texapon k 14, trombavar, natri tetradecylsulfas, sotradecol sodium, tetradecyl sodium sulfate
23665770
sodium;tetradecyl sulfate
[Na+].CCCCCCCCCCCCCCOS([O-])(=O)=O

Sodium 1-tetradecyl sulfate is used as an anionic surfactant in nickel plating bath, pickling, electrolytic cleaning, welding, leather dyeing and rubles polishes. It is used to study the time course of liquid drainage and foam coarsening and to investigate the effects of sclerosant foam preparation. It is used for the treatment of varicose and spider veins of the leg.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 1-tetradecyl sulfate is used as an anionic surfactant in nickel plating bath, pickling, electrolytic cleaning, welding, leather dyeing and rubles polishes. It is used to study the time course of liquid drainage and foam coarsening and to investigate the effects of sclerosant foam preparation. It is used for the treatment of varicose and spider veins of the leg.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 182°C to 192°C
Quantity 1 g
Beilstein 3578122
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 316.44
Percent Purity 95%
Assay cont. up to ≈5% sodium methyl sulfate
Chemical Name or Material Sodium 1-tetradecyl sulfate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'