Learn More
Scientific Glass Labs™ Soda Lime Glass Watch Glasses
Description
Watch glasses are useful for evaporating liquids, holding solids for weighing, or for heating small substances.
- Manufactured from Soda lime glass
- Thin, clear glass
- Ground edge
Specifications
Specifications
| Bottom Shape | Flat |
| Material | Soda Lime Glass |
| Shape | Round |
| Type | Watch Glass |
| For Use With (Application) | Evaporating Liquids, Heating Small Substances, Holding Solids for Weighing |
| Interior Finish | Ground Edge |
| Diameter (Metric) | 40 mm |