Learn More
BRAND™ Soda-Lime Glass Watch Glasses
Product Code. p-8004900 Shop All Brand Products
Change view
Click to view available options
Diameter (Metric):
50 mm
60 mm
70 mm
80 mm
90 mm
100 mm
125 mm
150 mm
200 mm
Unit Size:
Pack of 10
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10771572 125 mm Pack of 10
10313031 150 mm Pack of 10
10079980 60 mm Pack of 10
10273391 90 mm Pack of 10
10588831 200 mm Pack of 10
10558401 70 mm Pack of 10
10389901 100 mm Pack of 10
10455771 80 mm Pack of 10
15201489 50 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10771572 Supplier BRAND™ Supplier No. 150065
Only null left
Add to basket
Add to basket
Stress-reduced to minimize risk of shattering.
  • Soda-lime glass to DIN 12341
  • Ground edges
  • Stress-reduced to minimize risk of shattering
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DIN 12341
Bottom Shape Flat
Material Soda Lime Glass
Shape Round
Type Watch Glass
Interior Finish Polished
Diameter (Metric) 125 mm