Learn More
DWK Life Sciences Kimble Chase™ Soda Lime Glass Tube
Description
- Reusable
- 200Pack
- Clear soda lime glass
- With cap
Specifications
Specifications
| Length (Metric) | 40 mm |
| Packaging | 200Pack |
| Diameter (Metric) | 19 mm |
| Product Type | Test Tube |