Learn More
DWK Life Sciences Kimble Chase™ Soda Lime Glass Culture Test Tubes
Description
- Soda lime glass
- Round bottom
- X250
Specifications
Specifications
| Design Type | Round-bottom |
| Description | Test Tube |
| Length (Metric) | 75 mm |
| Material | Soda Lime Glass |
| Quantity | 250 Pack |
| Autoclavable | Autoclavable |
| Diameter (Metric) | 11.5 mm |