Learn More
Hirschmann™ Soda-lime Glass Test Tube with Round Bottoms
Product Code. 10438752
Change view
Click to view available options
Diameter (Metric):
12 mm
16 mm
Length (Metric):
75 mm
100 mm
125 mm
150 mm
160 mm
180 mm
Unit Size:
Pack of 100
Product Code. Diameter (Metric) Length (Metric) unitSize
10438752 16 mm 150 mm Pack of 100
10508211 12 mm 100 mm Pack of 100
10099730 16 mm 125 mm Pack of 100
10728621 16 mm 160 mm Pack of 100
10468942 16 mm 100 mm Pack of 100
10528771 12 mm 75 mm Pack of 100
13280500 16 mm 180 mm Pack of 100
7 options
Product Code. 10438752
Brand: Hirschmann™ 9270115
Only null left
Add to basket
Add to basket
Test tubes without scale made from sodalime glass.
  • Plain end
  • Round bottom
  • Sodalime-glass
Specifications
Length (Metric) 150 mm
Material Soda Lime Glass
Packaging 1Pack
Quantity 1 Pack
Diameter (Metric) 16 mm
Product Type Test Tube