Learn More
DWK Life Sciences Kimble Chase™ Soda Lime Glass Culture Tube
Product Code. 11882393
Change view
Click to view available options
Length (Metric):
40 mm
50 mm
55 mm
70 mm
120 mm
125 mm
150 mm
Diameter (Metric):
8 mm
9.5 mm
10.5 mm
11 mm
12 mm
16 mm
Unit Size:
Pack of 200
Pack of 250
Pack of 500
Product Code. Length (Metric) Diameter (Metric) unitSize
11882393 40 mm 10.5 mm Pack of 500
11892393 55 mm 11 mm Pack of 500
11842383 70 mm 11 mm Pack of 200
11803370 120 mm 12 mm Pack of 500
11862393 125 mm 16 mm Pack of 250
11832383 50 mm 9.5 mm Pack of 200
11822383 40 mm 8 mm Pack of 200
10364713 150 mm 16 mm Pack of 250
8 options
Product Code. 11882393
Brand: DWK Life Sciences A404010500911
Only null left
Add to basket
Add to basket
Use the Soda Lime Glass Culture Tube to view cell cultures in the laboratory. This disposable soda lime glass rim top tube is plain (unlabeled) and features a sturdy, uniform bottom.
  • Rimmed top
  • Unlableled
  • Clear soda lime glass
  • Uniform bottom
  • Disposable
TRUSTED_SUSTAINABILITY
Specifications
Design Type Round bottom
Length (Metric) 40 mm
Packaging 500Pack
Diameter (Metric) 10.5 mm
Product Type Test Tube