Description
- Soda Lime Glass
- Rimless
- Disposable
Specifications
Specifications
| Certifications/Compliance | ISO 4142 |
| Length (Metric) | 150 mm |
| Material | Soda Lime Glass |
| Quantity | 100 Pack |
| Rimmed | No |
| Diameter (Metric) | 16 mm |