Learn More
BRAND™ Soda Lime Glass Petri Dishes
Product Code. p-8011231 Shop All Brand Products
Change view
Click to view available options
Diameter (Metric):
40 mm
60 mm
80 mm
100 mm
150 mm
Height (Metric):
12 mm
15 mm
20 mm
25 mm
Unit Size:
Pack of 10
Product Code. Diameter (Metric) Height (Metric) unitSize
10624552 100 mm 15 mm Pack of 10
10400873 150 mm 25 mm Pack of 10
10019880 40 mm 12 mm Pack of 10
10450293 60 mm 15 mm Pack of 10
10315571 80 mm 15 mm Pack of 10
10039890 100 mm 20 mm Pack of 10
6 options
Product Code. 10624552
Brand: BRAND™ 455742
Only null left
Add to basket
Add to basket
Perfect for microbial or cell culture applications, these Soda Lime Glass Petri Dishes feature excellent glass quality and workmanship.
  • Dish and lid are flat inside and outside
  • Free from bubbles and streaks
  • Cut edges are fire-polished
  • Suitable for short-term chemical exposure
TRUSTED_SUSTAINABILITY
Specifications
Material Soda Lime Glass
Shape Round
Type Petri Dish
Edges Fire-polished cut edges
Height (Metric) 15 mm
Diameter (Metric) 100 mm