Learn More
Fisherbrand™ Soda Lime Glass Funnel
Product Code. 11562423 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric)Top:
45mm
50mm
55mm
60mm
75mm
100mm
Length (Metric) Stem:
45mm
50mm
55mm
60mm
70mm
100mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top Length (Metric) Stem Unit Size
11562423 100mm 100mm Each
11512423 45mm 45mm Each
11522423 50mm 50mm Each
11552423 75mm 70mm Each
11532423 55mm 55mm Each
11542423 60mm 60mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11562423
Estimated Shipment: 16-11-2026
Plain glass funnel manufactured from soda lime glass. Fisherbrand™ Soda Lime Glass Funnel is ideal for use with funneling liquids in laboratory applications.
Plain glass funnel manufactured from soda lime glass.
Specifications
Diameter (Metric)Top 100mm
Material Soda Lime Glass
Length (Metric) Stem 100mm