Learn More
Brand™ Soda Lime Glass Culture Tubes
Product Code. 30243952 Shop All Brand Products
Change view
Click to view available options
Length (Metric):
75 mm
100 mm
125 mm
160 mm
Diameter (Metric):
10 mm
12 mm
16 mm
Unit Size:
Pack of 100
Pack of 105
Pack of 144
Pack of 250
Pack of 344
Pack of 78
Product Code. Length (Metric) Diameter (Metric) unitSize
30243952 125 mm 16 mm Pack of 344
10261261 75 mm 10 mm Pack of 250
12209309 100 mm 16 mm Pack of 78
12088359 125 mm 16 mm Pack of 105
10211361 160 mm 16 mm Pack of 100
30243948 160 mm 16 mm Pack of 344
10411521 75 mm 12 mm Pack of 250
12626215 100 mm 12 mm Pack of 144
8 options
Product Code. 30243952
Brand: Brand™ 114120
Only null left
Add to basket
Add to basket
Manufactured from soda lime glass. BRAND™ Soda Lime Glass Culture Tubes are rimless and autoclavable at 121°C.
  • Manufactured from soda lime glass.
  • Rimless
  • Autoclavable at 121°C.
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 125 mm
Material Soda Lime Glass
Quantity 100/Pk.
Diameter (Metric) 16 mm
Product Type Culture Tube